Learn More
BRAND™ Tamper-Evident Screw Cap
Product Code. 10582774 Shop All Brand Products
Click to view available options
Color:
Blue
Green
Purple
Red
Transparent
Yellow
Unit Size:
Pack of 1000
Product Code. 10582774
Brand: BRAND™ 780771
Only null left
Add to basket
Add to basket
Tamper-Evident Screw Cap equipped with a silicone seal is suitable for use with microtubes.
  • With silicone seal
  • For microtubes
  • Working temperature between -196 to 121°C
TRUSTED_SUSTAINABILITY
Specifications
Includes Silicone Seal
Liner Material Silicone
Product Type Tamper-evident Screw Cap
Material Polypropylene
Color Blue
For Use With (Equipment) 78076064 Microtubes