Learn More
Applied Biosystems™ TaqMan™ Array Human Huntington Disease Pathway
Brand:  Applied Biosystems™ 4414140
Additional Details : Weight : 0.00450kg
Product Code. 10621186
  • 228.00€ / 1 set
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. TaqMan Assays (TaqMan probe and PCR primer sets) are considered the gold standard in gene expression analysis, enabling you to achieve high specificity and sensitivity and a broad dynamic range.
  • Extensive collections - choose from over 130 predesigned TaqMan™ Gene Signature Arrays and Plates, or customize your own plate, selecting from any of our 1.3 million inventoried assays across 24 species
  • Affordable - minimum order of only one gene signature plate or six custom plates
  • Convenient - dried TaqMan probe and primer sets supplied in 96-well plates; just add master mix and sample, and then run
  • Quick results - Fast format typically provides results in under an hour
Specifications
Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.
96-well plate
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
TaqMan™
Human
Dried Down
Standard
FAM
Neurobiology, Genetic Diseases, Signal Transduction
1 x 20 μL Reactions