missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TASP 0433864
GREENER_CHOICE

Product Code. p-7121005
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18741068

Brand: Tocris Bioscience™ 5362/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective mGlu2 receptor positive allosteric modulator

TASP 0433864 is a selective mGlu2 receptor positive allosteric modulator (EC50 values are 199 and 206 nM for human and rat receptors, respectively). Exhibits negligible activity against a range of neurotransmitter targets, except 5-HT2B and MAO-B (72 and 92% inhibition at 10 μM, respectively). Potentiates inhibitory effect of DCG IV (Cat. No. 0975) on excitatory postsynaptic potentials in hippocampal slices and ketamine- and methamphetamine-induced increases in locomotor activity in rats and mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S)-2-[[4-(1,1-Dimethylethyl)phenoxy]methyl]-2,3-dihydro-5-methylimidazo[2,1-b]oxazole-6-carboxamide
CAS 1431980-60-7
Synonym mGlu2 receptor positive allosteric modulator
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C18H23N3O3
Formula Weight 329.39
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.