missing translation for 'onlineSavingsMsg'
Learn More

TCEP Hydrochloride (0.5M in water), TRC

Product Code. 30415185
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
10mL
Product Code. Quantity unitSize
30415185 10 mL 10mL
30412912 1 mL -
2 options
This item is not returnable. View return policy

Product Code. 30415185

Brand: Toronto Research Chemicals TRCT01251510ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51805-45-9
Formula Weight 286.0373
InChI Formula InChI=1S/C9H15O6P.ClH/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15;/h1-6H2,(H,10,11)(H,12,13)(H,14,15);1H
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
Molecular Formula C9 H15 O6 P . Cl H
Molecular Weight (g/mol) 286.65
SMILES Cl.OC(=O)CCP(CCC(=O)O)CCC(=O)O
Synonym Tris (2-Carboxyethyl) Phosphine Hydrochloride (TCEP)
Concentration or Composition (by Analyte or Components) 0.5 M
Chemical Name or Material TCEP Hydrochloride

Specifications

Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.