Description
- See results clearly on the large backlit display.
- The alphanumeric keypad and function keys allow for a simplified user experience.
- On-screen prompts and soft keys will guide you through the operation.
- Results can be stored in the Micro SD card. Data output via built-in RS232 or optional interfaces including RS232/RS485/USB device, Ethernet and Bluetooth/WiFi.
- Analog Output and Discrete I/O with relays.
- Features common industrial modes and supports a 50,000 item library and 1,000 user profiles which can be downloaded/uploaded for easy set-up (with SD Card).
- GMP/GLP with time/date for result traceability.
Specifications
Specifications
| Connection Type | Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock |
| Display Type | Backlit LCD, 256 x 80 pixels |
| Dimensions (L x W x H) | 270 x 76 x 222 mm |
| Languages | English (US), French, German, Italian, Spanish (MX), Polish, Spanish |
| Weight (Metric) | 2.600 kg |
| Type | TD52XW-EU |