Learn More
OHAUS™ TD52XW Indicator
Product Code. 15938445 Shop All Ohaus Products
Change view
Click to view available options
Type:
TD52XW-EU
TD52XW-GB
Unit Size:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
15938445 TD52XW-GB 1 unit
15928445 TD52XW-EU 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15938445 Supplier OHAUS™ Supplier No. 30424067
Regulatory: This product is under eco-contribution tax.
OHAUS™ TD52XW Indicator ideal for use in conjunction with bench and floor scales for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
  • See results clearly on the large backlit display.
  • The alphanumeric keypad and function keys allow for a simplified user experience.
  • On-screen prompts and soft keys will guide you through the operation.
  • Results can be stored in the Micro SD card. Data output via built-in RS232 or optional interfaces including RS232/RS485/USB device, Ethernet and Bluetooth/WiFi.
  • Analog Output and Discrete I/O with relays.
  • Features common industrial modes and supports a 50,000 item library and 1,000 user profiles which can be downloaded/uploaded for easy set-up (with SD Card).
  • GMP/GLP with time/date for result traceability.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Display Type Backlit LCD, 256 x 80 pixels
Dimensions (L x W x H) 270 x 76 x 222 mm
Languages English (US), French, German, Italian, Spanish (MX), Polish, Spanish
Weight (Metric) 2.600 kg
Type TD52XW-GB