missing translation for 'onlineSavingsMsg'
Learn More

Cytiva TE 62 Transfer Unit

Product Code. 12994840 Shop All Cytiva Products
Click to view available options
Type:
Blotting System
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 12994840

Brand: Cytiva TE62

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Includes

Safety lid with power cables; 4 gel cassettes; 8 foam sponges, 3mm thick; 4 foam sponges, 6mm thick; 25 sheets of blotter paper. Replacement gel cassettes, electrode panels and foam sponges available.

Requires

Power supply, not included

Efficiently transfers proteins, DNA, or RNA from four standard polyacrylamide or agarose gels up to 15 x 21cm in size, or from 16 mini-gels

  • Hinged cassettes, molded of tough, glass-filled polyethersulfone for high durability, provide equal pressure across the gel
  • Cassettes load easily, with even contact between the gel and transfer membrane
  • Integral heat exchanger serves as a buffer cooling system
  • Safe, sturdy electrode panels lock automatically in place
  • Evenly spaced vertical platinum wire on panel and 10cm spacing between panels promote efficient transfer

TRUSTED_SUSTAINABILITY

Specifications

Color Black, Gray
Type Blotting System
Includes Safety lid with power cables, 4 gel cassettes, 8 Foam sponges 3mm thick, 4 Foam sponges 6mm thick, 25 sheets of blotter Paper
Temperature (English) Operating 39.2°F to 104°F
Temperature (Metric) Operating 4°C to 40°C
Width (English) 11.02 in.
Width (Metric) 28 cm
Height (English) 6.49 in.
Height (Metric) 16.5 cm
Wattage 200 w
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.