Learn More
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology, Fisher BioReagents™
Product Code. 10143393
Click to view available options
Quantity:
1 L
4 L
Packaging:
Nalgene Poly Bottle
PolyPac™
Unit Size:
1 L
4 L
Product Code. 10143393
Brand: Fisher BioReagents BP13384
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology
Tris–EDTA (TE) is routinely used for suspending nucleic acid samples.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tris-EDTA
Name Note 100X Solution
CAS 77-86-1, 60-00-4, 7732-18-5
Color Undesignated
Physical Form Liquid
Quantity 4 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Packaging PolyPac™
Protease Protease free
Grade Molecular Biology