missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Calibration Buffer pH 10.01, Hanna Instruments™
Click to view available options
Quantity:
20 mL
Specifications
Color | Colorless |
pH | 10.01 |
Physical Form | Liquid |
Quantity | 20 mL |
Solution Type | Buffer |
Formulation | Aqueous Buffer Solution |
Odor | Odorless |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'