Learn More
BRAND™ Technical Quality PFA Bottle
Product Code. 10111552 Shop All Brand Products
Click to view available options
Capacity (Metric):
100 mL
500 mL
Unit Size:
Each
Product Code. 10111552
Brand: BRAND™ 130482
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Technical Quality PFA Bottle includes an ETFE screw cap and narrow mouth.
  • With screw caps made of ETFE
  • These bottles contain a small fraction of reprocessed PFA from our production process
  • The physical properties and chemical resistance compare with high purity PFA
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Material PFA
Mouth Narrow
Closure Material ETFE
Closure Type Screw Cap