Learn More
BRAND™ Technical Quality PFA Bottle
Shop All Brand Products
Click to view available options
Capacity (Metric):
100 mL
500 mL
Unit Size:
Each
Description
- With screw caps made of ETFE
- These bottles contain a small fraction of reprocessed PFA from our production process
- The physical properties and chemical resistance compare with high purity PFA
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Material | PFA |
| Mouth | Narrow |
| Closure Material | ETFE |
| Closure Type | Screw Cap |