missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TEK (TIE2) [Y897S], GST Tag Recombinant Protein

Product Code. 15920293
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15920293

Brand: Invitrogen™ A30519

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This TEK (TIE2) Y897S recombinant human protein (771-end) was expressed in insect cells. TEK (TIE2) Y897S recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for TEK (TIE2) [Y897S] is NP_000450.2.

TEK (TIE2) Y897S receptor tyrosine kinase is expressed principally on vascular endothelium. Disrupting TEK Y897S function in mice results in embryonic lethality with defects in embryonic vasculature, suggesting a role in blood vessel maturation and maintenance. Angiopoietin-1 is a secreted growth factor that binds to and activates the TEK receptor tyrosine kinase. SHP2 and GRB2 are recruited to the activated TEK kinase domain and are part of the cellular responses that mediate TEK function. TEK expression is upregulated in the endothelium of vascular 'hot spots' in human breast cancer specimens. However, TEK is also overexpressed in areas of active angiogenesis in normal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q02763
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 150mM NaCl, 0.1mM EDTA, 0.1mM PMSF, 0.25mM DTT, 10mM glutathione and no preservative; pH 7.5
Gene ID (Entrez) 7010
Molecular Weight (g/mol) 66.7 kDa
Name Human TEK (TIE2) [Y897S], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias TIE2
Common Name TIE2 (TEK) (Y897S Mutant)
Gene Symbol TEK
KinaseFamily TIE Kinase Family
Protein Length 771-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 771-end
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Active, Catalytic, Mutant
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.