missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEKT4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562875
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RLDSANLWVL VDCILRDTSE DL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
TEKT4 Polyclonal antibody specifically detects TEKT4 in Human samples. It is validated for Immunohistochemistry.Specifications
TEKT4 | |
Polyclonal | |
Unconjugated | |
TEKT4 | |
1700010L19Rik; RGD1308075; Tek4; Tekt4; tektin 4; Tektin4; tektin-4; Testicular microtubules-related protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150483 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WW24 | |
TEKT4 | |
Recombinant protein corresponding to Human TEKT4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only