missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEKT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549713
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous level of total Tektin 3 protein.
TEKT3 Polyclonal antibody specifically detects TEKT3 in Human samples. It is validated for ELISA, Western Blot.Specifications
TEKT3 | |
Polyclonal | |
Unconjugated | |
TEKT3 | |
4933407G07Rik; OMC45; TEKT3; tektin 3; tektin-3; testicular microtubules-related protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64518 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BXF9 | |
TEKT3 | |
Fusion protein corresponding to C terminal 300 amino acids of human tektin 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |