missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Telmisartan EP Impurity E, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Telmisartan EP Impurity E
Specifications
Specifications
| Chemical Name or Material | 1-[(2'-Carboxybiphenyl-4-yl)methyl]-4-methyl-2-propyl-1H-benzimidazol-6-carboxylic Acid |
| CAS | 884330-12-5 |
| Analyte or Component Names | 1-[(2'-Carboxybiphenyl-4-yl)methyl]-4-methyl-2-propyl-1H-benzimidazol-6-carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H24 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Estradiol 3-Valerate, Telmisartan Imp. E (EP), 1-[(2'-Carboxy[1,1'-biphenyl]-4-yl)methyl]-4-methyl-2-propyl-1H-benzimidazole-6-carboxylic acid, 1-[(2'-Carboxybiphenyl-4-yl)methyl]-4-methyl-2-propyl-1H-benzimidazol-6-carboxylic Acid, 1H-Benzimidazole-6-carboxylic acid, 1-[(2'-carboxy[1,1'-biphenyl]-4-yl)methyl]-4-methyl-2-propyl-, Telmisartan Imp E (EP), Telmisartan Impurity E |
| SMILES | CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)c4ccccc4C(=O)O)C(=O)O |
| Molecular Weight (g/mol) | 428.49 |
| Quantity | 100 mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction