Learn More
TENM4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564980
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: PGYPKPDMDAM EPSYELIHTQ MKTQEWDNSK SILGVQCEVQ KQLKAFVTLE RFDQLYGSTI TSCQQAPKTK KFASSGSVFG KGVKFALKDG RVTTDIISVA NEDGRR Highest antigen sequence identity to the following orthologs - mouse 96%, rat 94%.
Involved in neural development, regulating the establishment of proper connectivity within the nervous system. Plays a role in the establishment of the anterior-posterior axis during gastrulation. Regulates the differentiation and cellular process formation of oligodendrocytes and myelination of small-diameter axons in the central nervous system (CNS) (PubMed:26188006). Promotes activation of focal adhesion kinase. May function as a cellular signal transducer.Specifications
TENM4 | |
Polyclonal | |
Unconjugated | |
TENM4 | |
KIAA1302; ODZ4; TNM4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26011 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6N022 | |
TENM4 | |
Recombit Human TENM4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only