Learn More
Tensin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563238
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VGSGPHPPDT QQPSPSKAFK PRFPGDQVVN GAGPELSTGP SPGSPTLDID QSIEQLNRLI LELDPTFEPI PTHMNALGSQ ANGSV Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 73%.
Tensin 3 Polyclonal antibody specifically detects Tensin 3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Tensin 3 | |
Polyclonal | |
Unconjugated | |
Tns3 | |
BC023928; F830010I22Rik; RGD1564174; TEM6; TENS1; TENS3; tensin 3; tensin3; tensin-3; tensin-3-like; tensin-like SH2 domain containing 1; tensin-like SH2 domain-containing 1; tensin-like SH2 domain-containing protein 1; thyroid specific PTB domain protein; TNS3; TPP; TSN3; tumor endothelial marker 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64759 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68CZ2 | |
Tns3 | |
Recombinant protein corresponding to Human Tensin 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only