missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TEPP 46

Product Code. p-200050989
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18055962

Brand: Tocris Bioscience™ 7809/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective pyruvate kinase M2 (PKM2) allosteric activator

TEPP 46 is a potent and selective allosteric activator of pyruvate kinase M2 (PKM2) (EC50 = 92 nM); it exhibits no effect on PKM1. TEPP 46 binds at the dimer-dimer interface of the PKM2 homotetramer, blocks its translocation into the nuclues and promotes a constitutively PKM2 active state. TEPP 46 impairs H1299 lung cancer cell proliferation under hypoxic conditions but not under normoxia in vitro. It inhibits growth of H1299 tumor xenografts in mice. TEPP 46 can reverse fructose-derived F1P inhibition of PKM2 and prevent fructose promoted hypoxia adaptation of intestinal cells. TEPP 46 reduces activation, proliferation, and cytokine production of both T helper 17 (Th17) and Th1 cells in vitro. It also inhibits experimental autoimmune encephalomyelitis (EAE) in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-[(3-Aminophenyl)methyl]-4,6-dihydro-4-methyl-2-(methylsulfinyl)-5H-thieno[2',3':4,5]pyrrolo[2,3-d]pyridazin-5-one
CAS 1221186-53-3
Quantity 10 mg
Target PKM2
Molecular Formula C17H16N4O2S2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.