missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbufoxon Sulfoxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
POA
Chemical Identifiers
| CAS | 56165-57-2 |
|---|---|
| Molecular Formula | C9 H21 O4 P S2 |
| Molecular Weight (g/mol) | 288.36 |
| Synonym | Phosphorothioic acid, S-[[(1,1-dimethylethyl)sulfinyl]methyl] O,O-diethyl ester, O,O-Diethyl S-(tert-butylsulfinyl)methyl phosphorothioate, Terbufos oxon sulfoxide, Terbufoxon sulfoxide |
| IUPAC Name | 2-(diethoxyphosphorylsulfanylmethylsulfinyl)-2-methylpropane |
| SMILES | CCOP(=O)(OCC)SCS(=O)C(C)(C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30135108
|
Toronto Research Chemicals
TRC-T109085-10MG |
10 mg |
POA
10mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
30138543
|
Toronto Research Chemicals
TRC-T109085-2.5MG |
2.5 mg |
POA
2.5mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
30008269
|
Toronto Research Chemicals
TRC-T109085-1MG |
1 mg |
POA
1mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
| 56165-57-2 | |
| 288.36 | |
| 2-(diethoxyphosphorylsulfanylmethylsulfinyl)-2-methylpropane |
| C9 H21 O4 P S2 | |
| Phosphorothioic acid, S-[[(1,1-dimethylethyl)sulfinyl]methyl] O,O-diethyl ester, O,O-Diethyl S-(tert-butylsulfinyl)methyl phosphorothioate, Terbufos oxon sulfoxide, Terbufoxon sulfoxide | |
| CCOP(=O)(OCC)SCS(=O)C(C)(C)C |
Specifications
| 56165-57-2 | |
| Phosphorothioic acid, S-[[(1,1-dimethylethyl)sulfinyl]methyl] O,O-diethyl ester, O,O-Diethyl S-(tert-butylsulfinyl)methyl phosphorothioate, Terbufos oxon sulfoxide, Terbufoxon sulfoxide | |
| CCOP(=O)(OCC)SCS(=O)C(C)(C)C | |
| 288.36 | |
| Terbufos-oxon-sulfoxide |
| C9 H21 O4 P S2 | |
| InChI=1S/C9H21O4PS2/c1-6-12-14(10,13-7-2)15-8-16(11)9(3,4)5/h6-8H2,1-5H3 | |
| 2-(diethoxyphosphorylsulfanylmethylsulfinyl)-2-methylpropane | |
| 288.0619 |
Safety and Handling
Recommended Storage : -20°C
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.