Learn More
3M™ Nomad™ Terra Heavy Traffic Entrance Matting 8150
Product Code. p-7213738
Click to view available options
Dimensions (L x W):
152 x 91 cm
610 x 122 cm
Unit Size:
1 roll
Each
Product Code. 17148720
Brand: 3M™ Nomad™ 7100116692
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For removing and hiding dirt in heavy traffic areas. Durable vinyl loops scrape, trap, and hide dirt & moisture, minimizing re-tracking into the building. Provides resistance to grease and oil. Backed mat can be used for indoor applications and protects floor surfaces.
  • Coiled vinyl scraper entrance mat designed for heavy-traffic applications
  • Open coil structure removes, traps, and hides dirt & moisture for easy cleaning
  • Matting for use indoors, outdoors, or in recessed wells of high traffic areas
  • Resistant to grease and oil
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 152 x 91 cm
For Use With (Application) Heavy traffic areas
Thickness (Metric) 14 mm
Color Gray
Length (Metric) 152 cm
Material Coiled Vinyl
Type Entrance Mat
Width (Metric) 91 cm