Learn More
3M™ Nomad™ Terra Medium Traffic Entrance Matting 6050
Product Code. 17158720
Click to view available options
Dimensions (L x W):
152 x 91.4 cm
610 x 91.4 cm
180 x 122 cm
610 x 122 cm
Color:
Black
Blue
Brown
Gray
Red
Unit Size:
1 roll
Each
Product Code. 17158720
Brand: 3M™ Nomad™ 7100116693
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This backed vinyl matting is suitable for indoor, medium traffic applications. Resilient vinyl loop construction traps, holds, and hides dirt/sand so surfaces stay neat and clean.
  • First step mat for efficient dirt removal
  • Suitable for medium traffic volumes
  • Collects and prevents dirt from being tracked into the building
  • Holds and hides dirt & sand, so the surface stays neat and clean
  • Foam backing reduces creeping and, together with the coils, provides additional cushioning for increased comfort
  • Easy installation
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 152 x 91.4 cm
For Use With (Application) Medium traffic areas
Thickness (Metric) 11 mm
Color Gray
Length (Metric) 152 cm
Material Vinyl
Type Entrance Mat
Width (Metric) 91.4 cm