missing translation for 'onlineSavingsMsg'
Learn More

Melford Terrific Broth, Granulated

Product Code. 30018828
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018828

Brand: Melford T151005000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enriched media for maintenance and propagation of E.

Enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant problem. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.

Ingredients:

Casein Digest Peptone: 12g/l

Yeast Extract: 24g/l

Dipotassium phosphate: 9.5g/l

Monopotassium phosphate: 2.2g/

Specifications

Quantity 5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.