missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ tert-Butyl propiolate, 98%

Brand:  Alfa Aesar™ H51048.06

Additional Details : CAS : 13831-03-3 Weight : 0.00500kg Transport : UN number : 3272 Chem class : 3 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15407937

  • 461.00€ / 5g


Estimated Shipment
14-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

13831-03-3
126.155
XGTPDIIFEPTULX-UHFFFAOYSA-N
543038
CC(C)(C)OC(=O)C#C
C7H10O2
MFCD00060100
tert-butyl propiolate, propiolic acid tert-butyl ester, t-butyl propiolate, 2-propynoic acid, 1,1-dimethylethyl ester, boc-acetylene, tert-butyl propalate, tert-butyl propargylate, acmc-209chj, tert-butyl propiolate #
tert-butyl prop-2-ynoate

Specifications

tert-Butyl propiolate
28°C (82°F)
Air Sensitive
18°C to 20°C
13831-03-3
HC≡CCO2C(CH3)3
UN3272
tert-butyl propiolate, propiolic acid tert-butyl ester, t-butyl propiolate, 2-propynoic acid, 1,1-dimethylethyl ester, boc-acetylene, tert-butyl propalate, tert-butyl propargylate, acmc-209chj, tert-butyl propiolate #
CC(C)(C)OC(=O)C#C
126.155
126.16
0.919
1.418
52°C to 53°C (27mmHg)
5g
C7H10O2
MFCD00060100
1747175
XGTPDIIFEPTULX-UHFFFAOYSA-N
tert-butyl prop-2-ynoate
543038
98%
Videos
SDS Documents