Learn More
TES Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553294
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QLPAHDQDPS KCHELSPREV KEMEQFVKKY KSEALGVGDV KLPCEMDAQG PKQMNIPGGD RSTPAAVGAM EDKSAEHKRT QYSCYCCKLS MKEGDPAIYA ERAGYDKLWH PACFVCSTCH ELLVD Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
TES Polyclonal antibody specifically detects TES in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TES | |
Polyclonal | |
Unconjugated | |
TES | |
D6Ertd352e; TES; Tes1; TES1/TES2; Tes2; TESS; TESS-2; testin; testin 2; testin LIM domain protein; testin2; testis derived transcript; testis derived transcript (3 LIM domains) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26136 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UGI8 | |
TES | |
Recombinant protein corresponding to Human TES | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only