Learn More
IKA™ Test Tube Attachment for IKA Vortex Shaker
Product Code. 10656263 Shop All IKA Products
Click to view available options
Holds:
12 x 12 mm Reagent Glasses
18 x 10 mm Reagent Glasses
8 x 16 mm Reagent Glasses
8 x 20 mm Reagent Glasses
Product Type:
Test Tube Adapter Attachment
Unit Size:
Each
Product Code. 10656263
Brand: IKA™ 0003344100
Only null left
Add to basket
Add to basket
Attachment with customized array for holding reagent glasses on a vortexed.
  • Requires VG 3.3 universal attachment
  • Facilitates multi-tube vortexing
TRUSTED_SUSTAINABILITY
Specifications
Holds 8 x 16 mm Reagent Glasses
Tube Size 16 mm
Product Type Test Tube Adapter Attachment
Description Test tube adapter attachment
Model Model VG 3.34
For Use With (Equipment) VG 3.3 Universal Attachment, Vortexer,
No. of Positions 8
Requires VG 3.3 Universal Attachment