Learn More
Velp Scientifica™ Test Tube Rack
Product Code. 15359564 Shop All Velp Products
Click to view available options
No. of Positions:
7
20
36
Holds:
20 17 mm Diamter Tubes
36 11 mm Diamter Tubes
7 29 mm dia. Tubes
Unit Size:
Each
Product Code. 15359564
Brand: Velp Scientifica™ A00000092
Only null left
Add to basket
Add to basket
Velp Scientifica™ Test Tube Rack is a fundamental accessory of the Velp Scientifica™ (OCB) Open Circulating Bath.
  • Completely transparent for a constant visual control
  • Supports variety of test tubes sizes
Specifications
Description Test Tube Rack
For Use With (Equipment) OCB
Holds 20 17 mm Diamter Tubes
Material Polycarbonate
No. of Positions 20