Learn More
Grant Instruments™ Test Tube Tray for Grant Shaking Water Baths
Click to view available options
Type:
Flask Tray
Test Tube Tray
Test Tube/Microtube Tray
For Use With (Equipment):
LSB12
LSB18
OLS26
Unit Size:
Each
Description
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruemnts™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
- Accommodates flasks from 25 to 1000mL
- For use with the followng Shaking Water Bath models:
- LSB12
- LSB18
- OLS26
- Compatible with SH plate holders or SR racks
- TF26 Test tube tray also compatible with SR racks
- Can be used alone to accommodate bags and miscellaneous vessels.
Specifications
Specifications
| Capacity (Metric) | 25 to 1000 mL |
| For Use With (Equipment) | OLS26 |
| Type | Flask Tray |