Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint, with Conical Bottom
Click to view available options
Diameter (Metric):
17 mm
Length (Metric):
115 mm
120 mm
Closure Type:
Hexagonal
Not Included
Stopper
Unit Size:
Pack of 10
Description
- With conical bottom
- With socket NS 14/23
- Available without stopper
- With PE stopper or with hexagonal hollow glass stopper
- Made of DURAN™ tubing
Specifications
Specifications
| Length (Metric) | 115 mm |
| Material | Borosilicate Glass (3.3) |
| Diameter (Metric) | 17 mm |
| Closure Type | Not Included |
| Bottom Shape | Conical |