missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydrogen Sulfate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073599
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
32503-27-8
C16H37NO4S
339.54
11637
SHFJWMWCIHQNCP-UHFFFAOYSA-M
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
94433
hydrogen sulfate;tetrabutylazanium
OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
This item is not returnable. View return policy
32503-27-8
C16H37NO4S
339.54
11637
SHFJWMWCIHQNCP-UHFFFAOYSA-M
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
94433
hydrogen sulfate;tetrabutylazanium
OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
Melting Point 168°C
Color White
pH 2.1
Packaging Plastic powder jar
Quantity 25 g
Formula Weight 339.53g/mol
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Tetrabutylammonium hydrogen sulfate
Signal Word
  • Danger||Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H315-Causes skin irritation.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'