missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl Methylidenebisphosphonate, TRC

Product Code. 30410785
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30410785 50 g 50g
30415574 5 g -
2 options
This item is not returnable. View return policy

Product Code. 30410785

Brand: Toronto Research Chemicals TRCT29280050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660-94-2
Formula Weight 288.0892
InChI Formula InChI=1S/C9H22O6P2/c1-5-12-16(10,13-6-2)9-17(11,14-7-3)15-8-4/h5-9H2,1-4H3
IUPAC Name 1-[diethoxyphosphorylmethyl(ethoxy)phosphoryl]oxyethane
Molecular Formula C9H22O6P2
Molecular Weight (g/mol) 288.21
SMILES CCOP(=O)(CP(=O)(OCC)OCC)OCC
Synonym P,P'-Methylenebisphosphonic Acid P,P,P',P'-Tetraethyl Ester, Methylenebisphosphonic Acid Tetraethyl Ester, Bis(diethylphosphono)methane, Methylenebis(diethoxyphosphine oxide), Methylenebis(diethyl phosphonate), Methylenebis(phosphonic acid) tetraethyl Ester, Tetraethyl methylenediphosphonate
Chemical Name or Material Tetraethyl Methylidenebisphosphonate

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.