Learn More
TEX19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562983
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PPVSMRYEEE GMSYLYASWM YQLQHGDQLS ICFTCFKAAF LDFKDLLESE DWEEDNWDPE LMEHTEAESE QE Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 61%.
TEX19 Polyclonal antibody specifically detects TEX19 in Human samples. It is validated for Immunohistochemistry.Specifications
TEX19 | |
Polyclonal | |
Unconjugated | |
TEX19 | |
FLJ35767; testis expressed 19; Testis-expressed protein 19; testis-expressed sequence 19 protein; TEX19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
400629 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NA77 | |
TEX19 | |
Recombinant protein corresponding to Human TEX19 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only