Learn More
Cytiva (formerly Pall Lab) TF PTFE Membrane Disc Filters
Strong, chemically resistant membranes for air monitoring and sampling in aggressive environments.
Brand:  Cytiva (formerly Pall Lab) 66155
Additional Details : Weight : 0.13300kg
 View more versions of this product
Product Code. 17124551
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Low chemical background permits highly sensitive, interference-free determinations
  • Ensures accurate gravimetric determinations with low tare mass
  • Ideal for filtration of gas and/or organic solvents
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
PTFE
1 μm
Non-sterile
47 mm