missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAP2D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567806
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse samples.
Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5′-GCCNNNGGC-3′ and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC.Specifications
TFAP2D | |
Polyclonal | |
Unconjugated | |
TFAP2D | |
TFAP2D; TFAP2BL1; Transcription factor AP-2-delta; Activating enhancer-binding protein 2-delta; Transcription factor AP-2-beta-like 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
226896, 83741 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7Z6R9, Q91ZK0 | |
TFAP2D | |
Synthesized peptide derived from human TFAP2D. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |