missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ebro Electronic™ TFI 250 Infrared Thermometers
Suitable for food and storage applications, kitchen inspections and food distribution. Ebro Electronic™ TFI 250 Infrared Thermometers come with adjustable sensitivity.
Brand: Ebro Electronic™ 13401753
Additional Details : Weight : 0.80000kg
Description
- Fast measurement
- Replaceable battery
- With laser pointer
- White, sturdy ABS plastic housing
Specifications
Factory calibration certificate | |
0.95 standard, adjustable from 0.1 to 1.0 | |
±0.1°C | |
550°C | |
-60°C to +550°C |
153 x 115 x 48 mm | |
TFI 250 | |
approx. 177 g | |
-60°C | |
±1.5°C |