Learn More
Ebro Electronic™ TFN 520 / 530 Robust Thermocouple Thermometer Series
Product Code. 10389282
Click to view available options
Product Type:
1-channel Thermometer
2-channel Thermometer
Probe Type:
Lemosa Connector
SMP Connector
IP Rating:
IP40
IP52
Unit Size:
Each
Récupel-bijdrage uitgesloten
Product Code. 10389282
Brand: Ebro Electronic™ 13405532A
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
High accurate thermometers for a whole range of measurement and control applications in the laboratory and in industry.
Temp. Range: -200 to 1200°C (K), -100 to 800°C (J), -50 to 300°C (T), -100 to 600°C (E)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012
Dimensions (L x W x H) 115 x 54 x 22 mm
Disposable Single-use
For Use With (Application) Applications: Heating technology, Air-conditioning, ventilation, Air, gases, Fluids, Motors, engines, Plastic materials, Laboratory
Height (Metric) 38 mm
Includes Thermometer, calibration certificate, instruction manual
Length (Metric) 190 mm
Material Plastic
Model TFN 530-SMP
Probe Type SMP Connector
Resolution 0.1°C at −200° to +250°C (K); 0.1°C at −100° to +190°C (J); 0.1°C at −50° to +220°C (T); 0.1°C at −100° to +150°C (E); 1°C for the rest
Weight (Metric) 0.3 kg
Product Type 2-channel Thermometer
Data Storage None
Max. Temperature (English) 2192°F
Max. Temperature (Metric) 1200°C
Min. Temperature (English) -328°F
Min. Temperature (Metric) -200°C
Temperature Range (Metric) -200°C to +1200°C
Calibrated Factory Calibrated
For Use With (Equipment) TPN probe series with SMP connector, AG 130, AG 140, AN 141, AN 150
Battery Life Up to 5 years
Battery Type Lithium 3 V/Typ CR 2477
Accuracy ±0.3°C
Display LCD
Width (Metric) 120 mm
IP Rating IP40