missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TGDS Recombinant Protein

Brand:  Abnova™ H00023483-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157362

  • 349.00€ / 10µg
Estimated Shipment: 10-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH33675
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
64.24
8
Glutathione Sepharose 4 Fast Flow
10 ug
MSAACWEEPWGLPGGFAKRVLVTGGAGFIASHMIVSLVEDYPNYMIINLDKLDYCASLKNLETISNKQNYKFIQGDICDSHFVKLLFETEKIDIVLHFAAQTHVDLSFVRAFEFTYVNVYGTHVLVSAAHEARVEKFIYVSTDEVYGGSLDKEFDESSPKQPTNPYASSKAAAECFVQSYWEQYKFPVVITRSSNVYGPHQYPEKVIPKFISLLQHNRKCCIHGSGLQTRNFLYATDVVEAFLTVLKKGKPGEIYNIGTNFEMSVVQLAKELIQLIKETNSESEMENWVDYVNDRPTNDMRYPMKSEKIHGLGWRPKVPWKEGIKKTIEWYRENFHNWKNVEKALEPFPV
SDR2E1/TDPGD
TGDS
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
23483
TGDS (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TGDS
Human
Recombinant
Solution
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions