missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallium Bromide, TRC
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
Specifications
Specifications
| Chemical Name or Material | Thallium Bromide |
| CAS | 7789-40-4 |
| Molecular Formula | BrTl |
| Quantity | 2.5 g |
| Synonym | Thallium Bromide (Tl2Br2), Thallium Bromide (TlBr)','Dimer, Thallium Monobromide, Thallium(I) Bromide, Thallous Bromide |
| InChI Formula | InChI=1S/BrH.Tl/h1H;/q;+1/p-1 |
| SMILES | [Br-].[Tl+] |
| IUPAC Name | thallium(1+);bromide |
| Molecular Weight (g/mol) | 284.29 |
| Formula Weight | 283.8927 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction