Learn More
DiscoverX Thapsigargin
Artikelnummer. 17302479
Encompass
missing translation for 'orderingAttributeHoverText'
:
1 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17302479
missing translation for 'mfr': DiscoverX 921148
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Thapsigargin is an inhibitor of SERCA. Provided as 1mg of dry powder.
Thapsigargin is a naturally occurring, cell-permeable plant sesquiterpene lactone that non-competitively inhibits the sarco/endoplasmic reticulum calcium ATPase (SERCA). Effectively increases intracellular calcium levels upon SERCA inhibition, leading to cell activation, increased histamine release from mast cells, and increased cancer cell proliferation. Acts as an anti-inflammatory and induces autophagy in-vivo. Provided as 1mg of dry powder.
Specifications
Quantity 1 mg