Learn More
Abeba™ The Original 5081 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 5081/42
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15671808
  • 57.67€ / 1 pair
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White/mint
  • Slip resistance outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Leather
Slip-resistant
Animal fats, oil and fuel
White
42
5081