Learn More
Abeba™ The Original 7204 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7204/43
Additional Details : Poids : 0.76300kg
 View more versions of this product
Product Code. 15662568
  • 38.20€ / 1 pair
Expédition estimée
28-09-2020
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather white with motif
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
  • Discontinued model
  • Heel cushion
Specifications
Specifications
Animal fats, oil and fuel
White
43
7204
CE EN ISO 20347:2012 OB FO SRC
Leather
Slip-resistant