Learn More
Kern Sohn Thermal Printer
Product Code. 17381753
Encompass_Preferred
Click to view available options
:
(L x W x H) 140 x 165 x 50 mm
Product Code. 17381753
Brand: Kern Sohn YKB01N
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermal printer for balances with data interface RS-232.
  • For printing weighing results, recipes, quantities etc. or for GLP record keeping of balance adjustment with date and time print
  • FLP/GMP compliant print form available as standard. Language versions: DE, GB, FR
Specifications
Connection Type Data Interface RS-232
Electrical Requirements Mains Adapter External, 100/240 V
Dimensions (L x W x H) 140 x 165 x 50 mm