Learn More
Thermo Scientific™ 384-Well Full-Skirted Plates, Standard
Click to view available options
Color:
Black
Unit Size:
Pack of 50
Description
Designed for high-throughput PCR, the Thermo Scientific 384-well plate is compatible with the majority of 384 block PCR and sequencing instruments. It has a maximum well volume of 40μL and a working volume of 25μL. This 384-well plate is compatible with all major 384 block thermal cyclers, and it has a cut corner at the A24 location.
Highlights
- Fully skirted for use with automated systems
- Compatible with all leading 384 block thermal cyclers
- 40μL maximum well volume, 25μL working volume
- Raised rim design around each well enables secure sealing and safeguards against evaporation
- Uniform, thin well walls deliver maximum and consistent heat transfer
- White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
- Cut corner: A24
Certifications
Certifed free from human DNA, RNases and DNases.
Specifications
Specifications
| Certifications/Compliance | Certifed free from human DNA, RNases and DNases |
| Format | Standard |
| Array | 16 x 24 |
| Color | Black |
| Material | Virgin Polypropylene |
| No. of Wells | 384 |
| Quantity | 50/Pk. |