Learn More
Thermo Scientific™ 4-Place Swinging Bucket Rotor Adapters
Product Code. 10759612 Shop All Thermo Scientific Products
Click to view available options
Tube Type:
15 mL Blood Tube
50 mL Falcon Tube
50 mL Tube
7 mL Tube
Falcon
Microtube
Unit Size:
Each
Product Code. 10759612
Brand: Thermo Scientific™ 75007542
For added processing versatility
  • Maximum tube size: 30 x 117mm
Order Info
Order 4-Place Swinging Bucket Rotor (75-007-591) with Round Buckets (75-007-555). Adapters are sold individually; order quantity to equip rotor. Note: 50mL conical adapter (75-007-556) requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
TRUSTED_SUSTAINABILITY
Specifications
Tube Type 50 mL Tube
For Use With (Equipment) Swinging bucket rotor