Learn More
Thermo Scientific™ 96-Well Separation Plate
Product Code. 11570304
Click to view available options
No. of Wells:
96
Unit Size:
Pack of 24
Product Code. 11570304
Brand: Thermo Scientific™ AB0952
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ready-to-use microplate format
  • 0.6mL well volume
  • Contains polyethylene frits
  • Pore size 30-45μm
  • Suitable for centrifugation
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
No. of Wells 96