Learn More
Thermo Scientific™ ART™ Non-Filtered Pipette Tips
Range of styles and packaging options
Brand:  Thermo Scientific™ 3920-11
Additional Details : Weight : 0.28350kg
 View more versions of this product
Product Code. 12612670
  • 19.79€ / Pack of 1000
Estimated Shipment
11-05-2021
Add to basket
Description
Description
Range of styles and packaging options
  • All sterile HLT branded tips are certified “Pure”
  • All nonsterile tips are manufactured in clean room environments and are certified DNA, RNase and DNase free
  • Stringent manufacturing and QA/QC testing procedures assure the most aseptic products on the market
  • Some tips have reference marks which provide a quick and convenient guide to visually establish whether you are pipeting the desired volume of liquid
Specifications
Specifications
200 μL
Yellow
Beveled
Non-sterile
Bulk