Description
Deionization System
For the Nanopure, E-pure Water Systems
Specifications
Specifications
| Pore Size | 60 μm |
| Resin Type | Mixed Bed |
| For Use With (Application) | Deionization System |
| For Use With (Equipment) | Quad, E-Pure, Deionization Cartridge |
| Type | Deionization Cartridge |