Learn More
Thermo Scientific™ Barnstead™ Pump Protectors for Water Purification Systems
Shop All Thermo Scientific ProductsDescription
- For installation in a pressurized feedwater line
- Shuts off pump if feedwater pressure falls below 5psi (0.35kg/cm2)
- Plugs into Nanopure (Nanopure Quad-style, Nanopure II or older Nanopure), E-pure, or older Easypure UV, UF, UV/UF and LF systems
- Can also be used with select Nanopure DIamond™ systems
- Includes everything necessary for installation
Specifications
Specifications
| For Use With (Equipment) | E-Pure Ultrapure Water Purification System |