Learn More
Thermo Scientific™ BioBasic™ 4 Javelin™ Guard Columns
Product Code. 11533291 Shop All Thermo Scientific Products
Click to view available options
:
Particle Size: 5μm; 20L x 1.0mm I.D.
Product Code. 11533291
Brand: Thermo Scientific™ 72305021006
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 20 mm
Diameter (Metric) Inner 1 mm
Stationary Phase BioBasic 4
Product Line BioBasic