Learn More
Fisherbrand™ Castaloy™ Chain Clamp
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
05-754 Q Clamp Holder
Unit Size:
Each
Description
For holding bottles, beakers and other large objects
- Holds 76 to 165mm (3 to 6.5 in.) bottles and large objects
- Fast, sure way to secure large equipment to rods and lattices
- Loop steel chain around object; attach to hook on movable slide; tighten slide with large, nonslip knob
- Holds equipment firmly in place at any angle, variable distance from support
- Arm is 15cm (6 in.) long and 13mm (0.5 in.) diameter
- Use with 05-754Q clamp holder
Specifications
Specifications
| Quantity | 1 Ea. |
| Type | Chain Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | 05-754 Q Clamp Holder |
| For Use With (Application) | For holding bottles, beakers and other large objects |
| Length (English) Arm | 6 in. |
| Length (Metric) Arm | 15.24 cm |