Learn More
Thermo Scientific™ CryoExtra™ Racks and Accessories
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack
Thermo Scientific CryoExtra 94 Rack
Type:
Cryo Mini Rack
Cryo Square Rack
Cryo Storage Frame
Holds:
2mL, 13 Cardboard Boxes (100-cell Divider)
2mL, 13 Cardboard Boxes (25-cell Divider)
2mL, 15 Cardboard Boxes (100-cell Divider)
2mL, 15 Cardboard Boxes (25-cell Divider)
Fenwal 4R5461 (250 mL Bag), Cassettes (5)
Fenwal 4R5462 (500 mL Bag), Cassettes (5)
Fenwal 4R9951 (50 mL Bag), Cassettes (8)
Unit Size:
Each
Specifications
Specifications
| Type | Cryo Storage Frame |
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | Thermo Scientific CryoExtra 94 Rack |
| Holds | Fenwal 4R9951 (50 mL Bag), Cassettes (8) |